missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein A Sepharose™ CL-4B Medium

For immunoglobin purification and fractionation. Cytiva Protein A Sepharose™ CL-4B Medium is also well suited for immunoprecipitation.

667.00 € - 19000.00 €

Specifications

Binding Capacity 20mg Human IgG/mL
Ligand Concentration 3mg Protein A/mL Medium
Matrix 4% Cross Linked Agarose
Particle Size 90 μm
pH 2 to 10
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11554955
View Documents
Cytiva
17-0963-02
500 mL
19000.00 €
500mL
In Stock
to see stock.
Add to basket
 
11514945
View Documents
Cytiva
17-0780-01
1.5 g
667.00 €
1500mg
In Stock
to see stock.
Add to basket
 
11564955
View Documents
Cytiva
17-0963-03
25 mL
2547.00 €
25mL
In Stock
to see stock.
Add to basket
 
Description

Description

  • Strong binding to Fc region of IgG
  • Suitable for polyclonal antibody purification
  • High capacity—20mg human IgG/mL
  • pH stability Cleaning-in-Place (CIP): 2 to 10
  • Stable in commonly used aqueous buffers, 6M guanidine-HCl, 8M urea

Specifications

Specifications

20mg Human IgG/mL
4% Cross Linked Agarose
2 to 10
For immunoglobin purification and fractionation and is suitable for immunoprecipitation procedures
3mg Protein A/mL Medium
90 μm
2°C to 8°C
Native Protein A
Videos
SDS
Documents

Documents

Product Certifications