Learn More
Protein C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA589751
346.50 EUR valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Description
Positive Samples: ES-2; Cellular Location: Endoplasmic reticulum, Golgi apparatus Immunogen sequence: ANSFLEELRH SSLERECIEE ICDFEEAKEI FQNVDDTLAF WSKHVDGDQC LVLPLEHPCA SLCCGHGTCI DGIGSFSCDC RSGWEGRFCQ REVSFLNCSL DNGGCTHYCL EEVGWRRCSC APGYKLGDDL LQCHPAVKFP CGRPWKRMEK KRSHLKRDTE DQEDQVDPRL IDGKMTRRGD SPWQVVLLDS KKKLACGAVL IHPSWVLTAA HCMDESKKLL VRLGEYDLRR WEKWELDLDI KEVFVHPNYS KSTTDNDI
Protein C is a vitamin K-dependent plasma glycoprotein that is cleaved to its activated form by the thrombin-thrombomodulin complex. Protein C in its activated form contains a serine protease domain and functions in degradation of the activated forms of coagulation factors V and VIII. Protein C is a vitamin K-dependent serine protease produced in the liver and made up of 2 polypeptide chains. The 62 kDa proenzyme is activated by thrombin bound to an endothelial surface receptor and the active enzyme cleaves factor Va and VIIIa and thus inhibits blood coagulation. The molecular weight of the active enzyme is 55 kDa and the normal concentrations in human plasma is approximately 1-3 ng/mL because of the very fast turnover, the proenzyme concentration is approximately 3 μg/mL. Mutations in the Protein C gene have been associated with thrombophilia due to protein C deficiency, neonatal purpura fulminans, and recurrent venous thrombosis.
Specifications
Protein C | |
Polyclonal | |
Unconjugated | |
PROC | |
Activation peptide, Anticoagulant protein C, APC, autoprothrombin IIA, Blood coagulation factor XIV, EDQVDPRLIDGK, inactivator of coagulation factors Va, VIII, PC, prepro-protein C, Proc, PROC1, protein C, protein C (inactivator of coagulation factors Va and VIIIa), protein C prepropeptide, protein C, inactivator of coagulation factors Va and VIIIa, THPH3, THPH4, type I protein C, vitamin K-dependent protein C, Vitamin K-dependent protein C heavy chain, Vitamin K-dependent protein C light chain | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19123, 5624 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.98 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
P04070, P33587 | |
PROC | |
Recombinant fusion protein containing a sequence corresponding to amino acids 43-300 of human Coagulation Protein C (NP_0003031) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.