missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Protein G Mag Sepharose™ Xtra
High binding capacity, 27mg human IgG/mL medium, for efficient capture of antibody with high yield and purity
124.81 € - 300.00 €
Specifications
Bead Type | Protein G Mag Sepharose Xtra magnetic bead |
---|---|
Binding Capacity | 27mg human IgG/mL gel |
Matrix | Paramagnetic, spherical, highly cross-linked agarose particles |
Content And Storage | 4°C to 8°C |
For Use With (Application) | For rapid, small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants |
Description
- Non adherant beads
- Matrix: paramagnetic, spherical, highly cross-linked agarose particles
- Ligand: Protein G
- Color: black
- Operating temperature: room temperature
- Can be used without detergents
- For small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants
- Available in 2 x 1mL and 5 x 1mL packs
Specifications
Protein G Mag Sepharose Xtra magnetic bead | |
Paramagnetic, spherical, highly cross-linked agarose particles | |
For rapid, small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants |
27mg human IgG/mL gel | |
4°C to 8°C | |
Protein G |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Protein G Mag Sepharose Xtra, 2 x 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-66.5 per cent of the mixture consists of component(s) of unknown acute toxicity.