missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein G Sepharose™ 4 Fast Flow Media

Rapid processing of large volumes, recoveries of 70 to 90%. Cytiva Protein G Sepharose™ 4 Fast Flow Media is recombinant protein G coupled to Sepharose 4 Fast Flow.

Brand:  Cytiva 17-0618-02

Code : GE

Additional Details : Weight : 0.07400kg


 View more versions of this product

Product Code. 11534935

  • 2686.00 € / 25mL
Estimated Shipment: 19-07-2024
to see stock.
Add to basket

Description

Description

  • Binding specificities that complement Protein A media
  • Binds a broad range of IgG species and subclasses
  • High capacity, >20mg human IgG/mL medium; high flowrate
  • Multi-point attachment minimizes ligand leakage
  • Used in a range of diagnostic and research applications

Specifications

Specifications

2mg Protein G/mL Medium
90 μm
25 mL
Used in a range of diagnostic and research applications
4% Cross Linked Agarose
3 to 9
2°C to 8°C
Recombinant Protein G lacking albumin binding region
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • protein G sepharose 4 fast flow

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers