missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control

For cellular and molecular biology applications

42.35 € - 53.00 €

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
For Use With (Application) A scrambled peptide with an identical amino acid composition to that of Protein Kinase C epsilon translocation Inhibitor Peptide. A cell impermeable negative control for PKC translocation inhibition.
Molecular Weight (g/mol) 843.98
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15430597
View Documents
Thermo Scientific Chemicals
J66521.!D
0.5 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15440597
View Documents
Thermo Scientific Chemicals
J66521.M^
1 mg
53.00 €
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A scrambled peptide with an identical amino acid composition to that of Protein Kinase C epsilon translocation Inhibitor Peptide. A cell impermeable negative control for PKC translocation inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
Solid
843.98
RUO - research use only
-30°C to -10°C
A scrambled peptide with an identical amino acid composition to that of Protein Kinase C epsilon translocation Inhibitor Peptide. A cell impermeable negative control for PKC translocation inhibition.
Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control
Leu-Ser-Glu-Thr-Lys-Pro-Ala-Val
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only