missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Protein and Peptide Molecular Weight Markers
Run these markers to determine the molecular weight of uncharacterized protein samples with precision. Cytiva Life Sciences™ Protein and Peptide Molecular Weight Markers work by comparing unknowns with carefully selected standards.
Brand: Cytiva 17-0445-01
345.57 EUR valid until 2025-12-31
Use promo code "24372" to get your promotional price.
This item is not returnable.
View return policy
Description
- Get clear, sharp bands for easy measurement of migration distances
- Bands can be visualized by using Coomassie™ Brilliant Blue or silver staining

Specifications
HMW Native Marker Kit | |
Protein and Peptide Molecular Weight Markers | |
For MW determinations by polyacrylamide gel electrophoresis (PAGE) in homogeneous or gradient gels. | |
Mr 669 000 | |
Unopened vials should be stored at 4°C |
White | |
Solid | |
Thyroglobulin: 76μg, 669000 (Mr), porcine thyroid (source), Ferritin: 50μg, 440000 (Mr), equine spleen (source), Catalase: 36μg, 232000 (Mr), bovine liver (source), Lactate dehydrogenase | |
10 Vials |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction