Learn More
Cytiva Whatman™ Protein Prep for ӒKTA™ Chromatography Systems
The Protein Prep syringe filter containing the regenerated cellulose membrane is the optimal choice for filtration prior to chromatography with ӒKTA™ systems.
Brand: Cytiva 10463033
475.84 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
0.45 μm | |
150 Pieces | |
30 mm |