missing translation for 'onlineSavingsMsg'
Learn More

PROTEIN S Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA590849

Product Code. 16558021

  • 472.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: MDINKPGPLF KPENGLLETK VYFAGFPRKV ESELIKPINP RLDGCIRSWN LMKQGASGIK EIIQEKQNKH CLVTVEKGSY YPGSGIAQFH IDYNNVSSAE GWHVNVTLNI RPSTGTGVML ALVSGNNTVP FAVSLVDSTS EKSQDILLSV ENTVIYRIQA LSLCSDQQSH LEFRVNRNNL ELSTPLKIET ISHEDLQRQL AVLDKAMKAK VATYLGGLPD VPFSATPVNA FYNGCMEVNI NGVQLDLDEA ISKHNDIRAH SCPSVWKKTK NS; Positive Samples: LO2, HeLa, 293T, Mouse liver, Mouse kidney, Rat liver, Rat kidney

Protein S is a vitamin K-dependent, potent neutral anticoagulant plasma glycoprotein. Protein S has a molecular weight of approximately 70 kDa and consists of a single polypeptide chain. The concentration of protein S in human plasma is 25 μg/mL, with a half-life of approximately two days. About 40% of Protein S in plasma is in a free form, whereas 60% is complexed with C4b-binding Protein (C4BP). Only the free Protein S functions as a cofactor to antigen presenting cells (APC) in the APC-dependent degradation of Factor Va and Factor VIIIa.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PROTEIN S
Polyclonal
Unconjugated
PROS1
AW214361, Preproprotein S, PROS, PROS 1, PROS1, protein S, protein S (alpha), Protein S alpha, protein Sa, proteinS, PS 21, PS 22, PS 23, PS 24, PS 25, PS 26, PS21, PS22, PS23, PS24, PS25, PS26, PSA, rabbit protein S, THPH5, THPH6, unnamed protein product, vitamin K-dependent plasma protein S, vitamin K-dependent protein S, vitamin K-dependent protein S precursor
Rabbit
Affinity Chromatography
RUO
19128, 5627, 81750
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
3.65 mg/mL
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3
P07225, P53813, Q08761
PROS1
Recombinant fusion protein containing a sequence corresponding to amino acids 405-676 of human PROS1 (NP_0003042)
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers