missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteinase K Lyophilized Powder, Thermo Scientific Chemicals
Brand: Thermo Scientific J76230.MC
This item is not returnable.
View return policy
Description
- Used in the isolation of native, high molecular weight DNA and RNA.
- Used as a general protease to inactivate DNases, RNases and to degrade proteins.
- Specifically modifies cell surface proteins and glycoproteins for analysis of membrane structures for protein localization.
- Produces characteristic protein fragments used in enzyme/protein structure and function studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Lyophilized powder
- Source: Tritirachium album
- Concentration: 20 - 50units/mg
- Molecular weight: 28.9kDa
- Optimum pH: 4.0 - 12.5
- Optimum temperature: 65°C
- Activators: Stimulated by 0.2 - 1% SDS or 1 - 4M urea
- Inactivated by PMSF(1); Not inactivated by EGTA, thiol reagents or trypsin/chymotrypsin inhibitors
Applications:
- Used in isolation of native, high molecular weight DNA and RNA
- Used as general protease to inactivate DNases, RNases and to degrade proteins
- Specifically modifies cell surface proteins and glycoproteins for analysis of membrane structures for protein localization
- Produces characteristic protein fragments used in enzyme/protein structure and function studies
Specifications
20 to 50 U/mg | |
Dry at 4°C | |
Proteinase K Powder | |
Lyophilized |
T. album | |
100 mg | |
28.9 kDa |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction