Learn More
R&D Systems™ Proteome Purify 12 Human Serum Protein Immunodepletion Resin
A single-use alternative to existing column or spin cartridge systems to deplete serum or plasma of high abundance proteins
Brand:  R&D Systems™ IDR012-040
 View more versions of this product
Product Code. 18472270
  • 1100.00 € / 40mL
Explore more special offers
Includes: Immunodepletion Resin, Spin-X Filter Units
Description
Description
Depletes samples of the 12 high-abundance proteins comprising approximately 95% of proteins in human serum or plasma
Depletes samples of albumin, IgA, IgG and IgM, Fibrinogen, Transferrin, Haptoglobin and others
Removal allows visualization of proteins
Specifications
Specifications
40 Tests
Immunodepletion Resin
Store the unopened product at 2 - 8° C. Do not use past expiration date.