missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel Bioanalysis™ Protino™ Ni-NTA Agarose
The method of choice for His-tag protein purification.
336.00 € - 4455.00 €
Specifications
Binding Capacity | < 50 mg/mL settled agarose |
---|---|
Description | Aqueous suspension |
Content And Storage | 4°C to 8°C |
For Use With (Application) | Purification of His-tag proteins |
Target | Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11912422
|
Macherey-Nagel Bioanalysis™
745400.25 |
25 mL |
336.00 €
25mL |
In Stock
Log in to see stock. |
|||||
11952192
|
Macherey-Nagel Bioanalysis™
745400.500 |
500 mL |
4455.00 €
500mL |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
Description
- Enable fast and convenient purification of recombinant polyhistidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).
- Proteins from any expression system can be purified under native or denaturing conditions.
- Binding of protein is based on the interaction between the polyhistidine tag of the recombinant protein and immobilized Ni2+ ions.
- Consists of the chelating ligand nitrilotriacetic acid (NTA) immobilized on 6% cross-linked agarose beads that are suitable for batch binding, gravity flow, and FPLC™ columns.
- The resin is precharged with Ni2+ ions and therefore ready to use.
- Technology: Immobilized metal ion affinity chromatography
- Batch binding, gravity flow column chromatography, MPLC / FPLC applications
- Matrix: 6 % beaded agarose (cross-linked), precharged with Ni2+
- Bead size: 45 to 165 μm
Specifications
< 50 mg/mL settled agarose | |
4°C to 8°C | |
Protein | |
NTA (nitrilotriacetic acid) |
Aqueous suspension | |
Purification of His-tag proteins | |
Agarose |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Flammable liquid Category 3
- Skin sensitiser Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : ethanol (diluted < 20 %) (denatured with1%IPA/1%MEK, acc.2016/1867/EU), resin suspension with Ni in
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title