missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protino™ Ni-TED Resin
For Purification of Polyhistidine-Tagged Proteins, Batch Binding, Gravity Flow Column Chromatography, MPLC/FPLC Applications
50.55 € - 1015.00 €
Specifications
Binding Capacity | 10mg/g |
---|---|
Binding Property | Batch |
Kit Contents | Buffers |
Matrix | Macroporous Silica |
Max. Operating Pressure | 145 psi |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11922532
|
Macherey-Nagel™
745200.5 |
5 g Dry Silica Bulk Resin |
50.55 €
5g |
In Stock
Log in to see stock. |
|||||
11962412
|
Macherey-Nagel™
745200.30 |
30 g Dry Silica Bulk Resin |
269.00 €
30g |
In Stock
Log in to see stock. |
|||||
11902292
|
Macherey-Nagel™
745200.120 |
120 g Dry Silica Bulk Resin |
1015.00 €
120g |
In Stock
Log in to see stock. |
|||||
Description
Protino™ NI-TED, the purer alternative for His-tag protein purification- Chelating group TED, one protein binding site
- High stability against reducing/chelating agents
- Low metal leaching
- High protein purity
- High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
- Dry resin, storage at room temperature
- Ready to use pre-packed columns filled with dry Protino™ Ni-TED resin
- Buffers included
- Dry bulk matrix
- Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink well - highest flexibility
- Batch procedure, gravity flow column chromatography, FPLC™ applications
- Empty Protino™ columns available for use with Protino™ Ni-TED resin
- Empty 96 well receiver plates available for use with Protino™ Ni-TED resin For purification of polyhistidine-tagged proteins; batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.
Specifications
10mg/g | |
Buffers | |
145 psi | |
Resin |
Batch | |
Macroporous Silica | |
For Purification of Polyhistidine-Tagged Proteins, Batch Binding, Gravity Flow Column Chromatography, MPLC/FPLC Applications | |
TED (tris-carboxymethyl ethylene diamine) |
Safety and Handling
Hazard Category
- Respiratory sensitiser Category 1
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title