Learn More
Invitrogen™ RNase-free Tips, 1000 μL size
Tips are RNase- and DNase-free and are provided in ten racks containing 96 tips each. They are compatible with Eppendorf, Finnpipette, Oxford Benchmate, Pipetman, Proline, Rainin, Socorex, and other brands of pipettors.
Brand:  Invitrogen™ AM12660TS
73.80 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 17769347
  • 123.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Ambion Tips are RNase- and DNase-free and are provided in ten racks containing 96 tips each
  • They are compatible with Eppendorf, Finnpipette, Oxford Benchmate, Pipetman, Proline, Rainin, Socorex, and other brands of pipettors
  • Each lot of tips is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 μL
Sterile
RNase-free Tips
Room Temperature
Ambion
Non-filtered
10 racks
Includes 10 racks with 96 tips each.
Store at room temperature.
Universal Fit