Learn More
PRPSAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17814-1-AP-150UL
 View more versions of this product
Product Code. 16827804
  • 410.00 € / 150µL
Explore more special offers
Description
Description
The enzyme phosphoribosylpyrophosphate synthetase (PRS) catalyzes the formation of phosphoribosylpyrophosphate which is a substrate for synthesis of purine and pyrimidine nucleotides, histidine, tryptophan and NAD. PRS exists as a complex with two catalytic subunits and two associated subunits. This gene encodes a non-catalytic associated subunit of PRS.
Specifications
Specifications
PRPSAP2
Polyclonal
Unconjugated
PRPSAP2
PAP41, PRPSAP2
Rabbit
Antigen Affinity Chromatography
RUO
117272, 212627, 5636
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08618, O60256, Q8R574
PRPSAP2
PRPSAP2 Fusion Protein Ag12132
150 μL
Primary
Human, Mouse, Rat