Learn More
Tocris Bioscience™ PSB 1114
Potent, selective P2Y2 agonist
Brand:  Tocris Bioscience™ 4333/500U
281.54 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18045402
  • 348.00 € / 500µg
Explore more special offers
Description
Description
PSB 1114 is a potent, selective P2Y2 agonist (EC50 = 0.134 μM). Displays >60-fold selectivity versus P2Y4 and P2Y6 receptors.
Specifications
Specifications
4-Thiouridine-5'-O-(β,γ-difluoromethylene)triphosphate tetrasodium salt
500 μg
C10H11F2N2Na4O13P3S
1657025-60-9
P2Y Receptor Agonists
98%