Learn More
PSF Progressive Safety™ PSF Strata Black Chukka Cut Resistant Boot
Reinforced interlayer lining to upper and tongue for excellent cut resistance
Brand:  PSF Progressive Safety™ 527SM-12
 View more versions of this product
Product Code. 12976776
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Full grain, water resistant leather upper
  • PU/rubber outsole
  • Steel midsole
  • Antistatic
  • Heat resistant to 300°C
Specifications
Specifications
12