Learn More
PSI-6206 13C,d3, MedChemExpress
MedChemExpress PSI-6206 13CD3 is the deuterium labeled PSI-6206. PSI-6206 is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.
Brand: Medchem Express HY-15236S
Description
PSI-6206 13CD3 is the deuterium labeled PSI-6206. PSI-6206 is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.Specifications
PSI-6206 13C,d3 | |
5 mg | |
Research | |
RO-2433 13C,d3 GS-331007 13C,d3 Sofosbuvir metabolite GS-331007 13C,d3 | |
264.23 | |
98.0% | |
Solid |
1256490-42-2 | |
Light Yellow | |
C9 13CH10D3FN2O5 | |
OC[C@@H]1[C@H]([C@@]([13C]([2H])([2H])[2H])(F)[C@H](N2C(NC(C=C2)=O)=O)O1)O | |
264.23 | |
Research |