missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMG3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15601-1-AP-150UL
This item is not returnable.
View return policy
Description
PSMG3, also named as C7orf48 and PAC3, is a chaperone protein which promotes assembly of the 20S proteasome. It may cooperate with PSMG1-PSMG2 heterodimers to orchestrate the correct assembly of proteasomes.Specifications
PSMG3 | |
Polyclonal | |
Unconjugated | |
PSMG3 | |
C7orf48, hPAC3, PAC 3, PAC3, PSMG3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100361517, 66506, 84262 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BT73, Q9CZH3 | |
Psmg3 | |
PSMG3 Fusion Protein Ag7984 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction