Learn More
Masterflex™ PTFE 3-Way Direct Lift Solenoid Valve
Brand:  Masterflex™ 01540-12
 View more versions of this product
Product Code. 12843223
  • 217.00 € / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Ideal for use with high-purity or corrosive fluids
PTFE
3-way Direct Lift Solenoid Valve
30 psi
70°C
1 Each
24 VDC