missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ PTFE Ultrapure Barrel Pump with Discharge Hose
Provides high chemical resistance and safe transfer, even with liquids which attack conventional materials such as PP or PVC.
Brand: Buerkle™ 5606-0401
This item is not returnable.
View return policy
Description
- Medium-contacting parts manufactured from PTFE
- Suitable for transferring ultra-pure liquids
- Stopcock prevents hazardous vapours from escaping and stops dripping after transfer
- Discharge hose is 1.2m in length
- Immersion tube with a diameter of 32mm
- All valves are made of solid PTFE/PVDF
- Top quality, 3 years’ warranty
Specifications
Immersion depth of 40 cm | |
40 cm | |
PTFE | |
Barrel Pump |
Discharge Tube | |
1 Ea. | |
180 mL/stroke | |
For transferring ultra-pure liquids |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Buerkle PTFE Ultrapure Barrel Pump with Discharge Hose
Topsellers fill off liquids
File size: 634.4 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buerkle™ PTFE Ultrapure Barrel Pump with Discharge Hose > Flow Rate: 180mL/stroke
Spot an opportunity for improvement?Share a Content Correction