missing translation for 'onlineSavingsMsg'
Learn More
PTFE Beaker Cover-Watch Glass Product Code.: 15486175
In Stock
to see stock.
Add to basket

PTFE Beaker Cover-Watch Glass

Product Code. 15486175
Watch glass; Fits 1000mL Beakers
Click to view available options
Description:
Watch glass; Fits 1000mL Beakers
Watch glass; Fits 100mL Beakers
Watch glass; Fits 2000mL Beakers
Watch glass; Fits 250mL Beakers
Watch glass; Fits 25mL Beakers
Watch glass; Fits 5-10mL Beakers
Watch glass; Fits 500mL Beakers
Watch glass; Fits 50mL Beakers
This item is not returnable. View return policy

Product Code. 15486175

Brand: Thermo Scientific Alfa Aesar 038098.KT

In Stock
to see stock.
Add to basket
Add to basket
Request Bulk or Custom Format
This item is not returnable. View return policy

For use as a beaker cover or a watch glass

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Available in a range of diameters
TRUSTED_SUSTAINABILITY

Specifications

Description Watch glass; Fits 1000mL Beakers
Material PTFE
Type Beaker

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PTFE Beaker Cover-Watch Glass >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.