missing translation for 'onlineSavingsMsg'
Learn More

PTFE Beaker Lids

Product Code. 15471477
Click to view available options
For Use With (Equipment):
1000mL beaker
100mL beaker
10mL beaker
2000mL beaker
250mL beaker
25mL beaker
500mL beaker
50mL beaker
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with beakers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Available in a range of diameters
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Outer 32mm
Material PTFE
For Use With (Equipment) 25mL beaker
Height (Metric) 47mm

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'