missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTFE Beaker Lids
Click to view available options
For Use With (Equipment):
1000mL beaker
100mL beaker
10mL beaker
2000mL beaker
250mL beaker
25mL beaker
500mL beaker
50mL beaker
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Available in a range of diameters
Specifications
Specifications
Diameter (Metric) Outer | 43mm |
Material | PTFE |
For Use With (Equipment) | 50mL beaker |
Height (Metric) | 60mm |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'