missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ PTFE Screw-top Bottle

Product Code. 10074340
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
150 mL
500 mL
1000 mL
2000 mL
Diameter (Metric):
22 mm
26 mm
33 mm
43 mm
52 mm
60 mm
80 mm
100 mm
120 mm
Diameter (Metric) Inner Neck:
10mm
12mm
19mm
25mm
35mm
52mm
57mm
60mm

Molded-in hand grips provide ease of manipulation. The PTFE screw-top bottle is chemically resistant and inert.

  • Isostatically molded in pure PTFE and with super smooth internal finish, FDA approved
  • Insoluble and non-contaminating
  • Exceptional thermal resistance can be used from -200°C to +280°C
  • Hand grips for ease of manipulation
  • Do not use, when sealed, for chemical reactions or digestions, loosen lid when thermal sterilizing

Specifications

Capacity (Metric) 10 mL
Color White
Diameter (Metric) Inner Neck 12mm
For Use With (Application) Storage of hazardous chemicals
Height (Metric) With Closure 50mm
Material PTFE
Max. Temperature (Metric) 280°C
Temperature Range (Metric) -200 to +280°C
Thickness (Metric) 2.25mm
Weight (Metric) 27g
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Closure Included Yes
Autoclavable Autoclavable
Graduated No
Product Type Storage Bottle
Diameter (Metric) 26 mm
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'