missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Bottle Pourer

Fisherbrand™ PTFE Bottle Pourer permits direct pouring of all liquids from reagent bottles in a safe and consistent manner.

57.10 €

Specifications

Description Fisherbrand™ PTFE Bottle Pourer
Material PTFE
For Use With (Equipment) Reagent Bottles
View More Specs

Products 4
Product Code Brand Neck ID (Metric) Price Quantity & Availability  
Product Code Brand Neck ID (Metric) Price Quantity & Availability  
16216031
View Documents Promotion Details
Fisherbrand™
028.030
GL30
57.10 €
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
 
12681912
View Documents Promotion Details
Fisherbrand™
028.045
57.10 €
Each
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
16226031
View Documents Promotion Details
Fisherbrand™
028.032
57.10 €
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
 
16236031
View Documents Promotion Details
Fisherbrand™
028.038
GL38
57.10 €
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
 
Description

Description

  • Totally inert with PTFE encapsulated viton neck seal
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications

Specifications

Fisherbrand™ PTFE Bottle Pourer
PTFE
Reagent Bottles
Videos
SDS
Documents

Documents

Product Certifications