missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ PTFE-Coated Micro Double Spatulas

Well-suited for small quantity applications and narrow-format tubes, these double-ended, PTFE-coated spatulas reduce material sticking. Bochem™ PTFE-Coated Micro Double Spatulas are also gentle on delicate, scratchable surfaces.

Brand:  Bochem™ 3710

 View more versions of this product

Product Code. 10202111

  • 27.30 € / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 18/10 stainless steel
  • Double endet
  • PTFE coating
Specifications

Specifications

1 Ea.
Double Ended Micro Spatula
3 mm
1.5 mm
150 mm
PTFE
40 mm
3 mm
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers