missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ PTFE-Coated Micro Double Spatulas

Well-suited for small quantity applications and narrow-format tubes, these double-ended, PTFE-coated spatulas reduce material sticking. Bochem™ PTFE-Coated Micro Double Spatulas are also gentle on delicate, scratchable surfaces.

Brand:  Bochem™ 3713

 View more versions of this product

Product Code. 10458442

  • 27.95 € / Each
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Non-stick PTFE coating.
  • Double-ended for versatility.
  • Slim ends for easy container access.
  • One rounded end and one flat end.
Specifications

Specifications

1 Ea.
Double Ended Micro Spatula
6 mm
210 x 6 mm
210 mm
PTFE
55 mm
2mm
PTFE Coating
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.