Learn More
Fisherbrand™ PTFE Crucibles
Guaranteed inert and crack-free. Fisherbrand™ PTFE Crucibles can be used up to 280°C.
13.05 € - 41.85 €
Specifications
Material PTFE
Product Type Specs Containers
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
16235941
View Documents
Fisherbrand™
009.005
5 mL
41.85 €
Pack of 5
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
10416212
View Documents
Fisherbrand™
009.025
25 mL
13.05 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
10119811
View Documents
Fisherbrand™
009.075
75 mL
18.55 €
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
Description
Description
  • Isostatically molded in pure PTFE
  • Heavy wall construction
Specifications
Specifications
PTFE
Containers