Learn More
Bohlender™ PTFE Cylindrical Magnetic Stirring Bar
Brand:  Bohlender™ C350-08
 View more versions of this product
Product Code. 10490194
  • 23.85 € / Pack of 10
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Specifications
FDA Certified
Alnico 5 magnetic core
10 mm
1/Pk.
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics
3 mm
PTFE
Cylinder