missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Dipper Beaker with Handle

Uses 600mm steel core within shaft for excellent rigidity. The PTFE dipper beaker is chemically resistant and inert.

Brand:  Fisherbrand™ 036.250

 View more versions of this product

Product Code. 10369690

  • 106.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

PTFE sampling container with a detachable 600mm long shaft.
  • Chemically resistant and inert
  • Shafts have a steel core for rigidity
  • Fully encapsulated in PTFE
  • Suitable for sampling the most aggressive media
Specifications

Specifications

Dipper Beaker
Autoclavable
PTFE
White
Water analysis, Sampling of general liquids including liquid nitrogen
-200°C to +280°C
67 mm (Bowl)
600 mm
PTFE
PTFE Dipper Beaker & PTFE Shaft
250 mL
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE Additional Shaft for PTFE Dipper Beakers
67 x 67 x 696 mm
93 mm
67 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.