Learn More
Fisherbrand™ Dipper Beaker Shafts
Suitable for sampling the most aggressive media. Fisherbrand™ Dipper Beaker Shafts are fully encapsulated in PTFE plastic for rigidity.
Brand:  Fisherbrand™ 036.001
Product Code. 15132569
  • 58.05 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Shafts have a steel core
  • Totally chemically resistant and inert
  • Additional shafts available to extend the dippers
Specifications
Specifications
Dipper Beaker
Autoclavable
White
PTFE Dipper Beakers
12 x 12 mm
600 mm
PTFE
PTFE
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
-200°C to +280°C
12 x 12 x 600 mm
12 mm