Learn More
Kartell™ Lining for Glass Tapers
Constructed of PTFE for high temperature and chemical resistance. Kartell™ Lining for Glass Tapers are specially designed to prevent machined glass joints from binding.
Brand:  Kartell™ 0104000
 View more versions of this product
Product Code. 10707741
  • 37.70 € / Pack of 10
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to prevent machined glass joints from binding
  • Conforms to DIN 12242
  • Suitable for foodstuff
Specifications
Specifications
DIN 12242
21 mm
Prevent glass joints from binding
24/29 Cone Size
PTFE
Glass Joint
10/Pk.