missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Disposable Ground Joint Sleeves, PTFE

Inserts between standard ground glass joints to eliminate the use of grease. Lenz Laborglasinstrumente™ Disposable Ground Joint Sleeves, PTFE provide outstanding chemical resistance at operating temperatures from -200 to +260°.

Brand:  Lenz Laborglasinstrumente™ 01101529

 View more versions of this product

Product Code. 10541775

  • 71.00 € / Pack of 10
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 50μm thickness
  • Simplifies use of joint clips
  • Fits like a second skin without adding bulk
Specifications

Specifications

PTFE
10Pack
NS 29/32
Ground Joint Sleeve
–200 to 260°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.