Learn More
DWK Life Sciences PTFE Hose Connection, with silicone seal (VMQ), complete with screw cap PBT, bent
For safe connection of hoses.
108.00 € - 129.00 €
Specifications
Quantity 10/Pk.
Type Bent
View More Specs
Products 2
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
10795295
View Documents
DWK Life Sciences
286230605
GL 14
108.00 €
Pack of 10
In Stock
to see stock.
Add to basket
10030704
View Documents
DWK Life Sciences
286230802
GL 18
129.00 €
Pack of 10
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
Description
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PTFE for use with higher thermal and chemical resistance requirements.
  • Hose olives enable the connection of hoses to laboratory apparatuses
Specifications
Specifications
10/Pk.
Bent