Learn More
DWK Life Sciences PTFE Hose Connection, with silicone seal (VMQ), complete with screw cap PBT, straight
For safe connection of hoses.
78.35 € - 164.00 €
Specifications
Quantity 10/Pk.
Type Straight
View More Specs
Products 3
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
10376272
View Documents
DWK Life Sciences
286230005
GL 14
78.35 €
Pack of 10
In Stock
to see stock.
Add to basket
10561293
View Documents
DWK Life Sciences
286230202
GL 18
90.45 €
Pack of 10
In Stock
to see stock.
Add to basket
15273428
View Documents
DWK Life Sciences
286230408
GL 25
164.00 €
Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PTFE for use with higher thermal and chemical resistance requirements.
  • Hose olives enable the connection of hoses to laboratory apparatuses
Specifications
Specifications
10/Pk.
Straight