Learn More
Quickfit™ PTFE Joint Clips with Conical Joints
Click to view available options
Size:
10/19
12/21
19/26
24/29
34/35
40/38
45/40
Unit Size:
Pack of 5
Description
Highly resistant to chemical attack
- Used for retaining glassware having Quickfit™ conical joints
- Manufactured from PTFE with encapsulated steel spring to provide firm grip
- Clips are numbered with joint size for easy identification
- Can be used at temperatures up to 250°C
Specifications
Specifications
| Type | Joint Clip |
| Material | PTFE |
| Includes | Encapsulated Steel spring |
| For Use With (Equipment) | Conical Joints |
| Size | 12/21 |
| Temperature (Metric) Resistant | 250°C |