Learn More
Quickfit™ PTFE Joint Clips with Conical Joints
Highly resistant to chemical attack
Brand:  Quickfit GC10
 View more versions of this product
Product Code. 12089052
  • 44.00 € / Pack of 5
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Highly resistant to chemical attack
  • Used for retaining glassware having Quickfit™ conical joints
  • Manufactured from PTFE with encapsulated steel spring to provide firm grip
  • Clips are numbered with joint size for easy identification
  • Can be used at temperatures up to 250°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Joint Clip
Encapsulated Steel spring
10/19
PTFE
Conical Joints
250°C