Learn More
DWK Life Sciences PTFE Key with Thread, with three part retaining device
For use with polished DURAN™ disposable taps.
Brand:  DWK Life Sciences 286221009
 View more versions of this product
Product Code. 15874618
  • 366.00 € / Pack of 25
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Disposable PTFE valve plugs in combination with the polished DURAN™ disposable valve represent a grease-free alternative. The 3-part tap safety device is already included in the scope of delivery.
  • Made of PTFE with thread and 3-part valve locking device
  • For use with polished DURAN™ one-way taps
  • Grease-free working
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6 mm
Not Autoclavable
25/Pk.
PTFE
18.8 NS
For use with polished DURAN™ one-way taps