Learn More
Cowie Technology™ PTFE Plain Stirring Magnets
Gives turbulence at low speeds.
Brand:  Cowie Technology™ 001.230.7
Product Code. 16205791
  • 14.30 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Gives turbulence at low speeds.
  • Gives turbulence at low speeds
Specifications
Specifications
White
30mm x 7mm
7mm
30 mm
Cylinder
Pure PTFE Magnet stir bar
Stirrers, hotplate stirrers
7 mm
1/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cowie Technology™ PTFE Plain Stirring Magnets > PTFE Plain Stirring Magnets