Learn More
Bohlender™ PTFE Plugs
Brand:  Bohlender™ D549-18
Product Code. 12770706
  • 23.00 € / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
FDA Conform
PTFE
-200°C
GL 18
Plug
For screw caps
1 Each
250°C