Learn More
Poulten Graf™ Fortuna™ Optifix™ Basic Dispenser
Poulten Graf™ Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.
Brand:  Poulten Graf™ 101 08056
 View more versions of this product
Product Code. 10798941
  • 2430.00 € / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For dosing of the most common chemicals, acids and crytallizing liquids (not suitable for solvents). Poulten Graf™ PTFE/PP/Fiberglass/Teflon/Ceramic Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.
  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • With solid PTFE encased dispensing piston made of glass
  • Cylinder coated with plastic to protect against breakage
  • With certificate, individually numbered
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment
Specifications
Specifications
Fortuna Optifix Basic Dispenser
PTFE, Polypropylene, Fiberglass, Teflon, Ceramic
For dispensing aqueous solutions, acids and crystallizing liquids
Clear
60 to 300 mL