Learn More
Fisherbrand™ Right Angle Connectors
For use with rigid and semi-rigid tubing consisting of glass, FEP or PTFE plastic. Fisherbrand™ Right Angle Connectors feature all wetted parts manufactured from pure PTFE plastic.
Brand:  Fisherbrand™ 016.1004.2
Product Code. 10212002
  • 13.40 € / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Bayonet fitting
  • New, improved connector system
Order Info
dimProductType
Specifications
Specifications
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Autoclavable
PTFE
Flexible tubing in PTFE, Viton, silicone rubber or similar materials
1 Each
16 mm
280°C
PTFE Right Angle Connector
10 mm
34 mm
-200°C
33.5 mm
White