missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Stir Bars with Pivot Ring

Product Code. 11868862 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
8 mm
12 mm
13 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
45 mm
50 mm
60 mm
70 mm
75 mm
Diameter (Metric):
3 mm
4.5 mm
6 mm
8 mm
10 mm
13 mm
Unit Size:
Each
Pack of 10
Pack of 5

Product Code. 11868862

Only null left
Add to basket
Edge
Add to basket

Provides stable stirring action. Fisherbrand™ PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.

Provides stable stirring action. FisherbrandTM PTFE Stir Bars with Pivot Ring are ideal for containers with uneven or unusual bottom surfaces and shapes.

  • Stable, general-purpose magnetic stir bar
  • The flat surfaces provide a greater mixing action.
  • Autoclavable

Specifications

Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core; Cylindrical with Pivot Ring
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Coating PTFE
Diameter (Metric) 6 mm
Length (Metric) 35 mm
Material PTFE
Shape Cylinder, Pivot ring
Type Stir Bar
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.