Learn More
Cowie Technology™ PTFE Straight Through Connector
Accepts any rigid or semi-rigid tubing. Cowie Technology™ PTFE Straight Through Connector locks in place quickly with a bayonet type coupling.
Brand:  Cowie Technology™ 016.905.2
 View more versions of this product
Product Code. 10038201
  • 11.35 € / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Bayonet fitting
  • For use with rigid and semi rigid tubing such as glass, FEP or PTFE
Specifications
Specifications
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Autoclavable
PTFE
-200°C
16 mm
White
PTFE Straight Through Connector
11 mm
54 mm
1 Each
16 mm
280°C