Learn More
Hirschmann™ PTFE Thread Adapter
Threaded adapter PTFE for use with ceramus™ -classic.
Brand:  Hirschmann™ 9313606
 View more versions of this product
Product Code. 10044310
  • 89.05 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For the adaption on the most common bottles
Specifications
Specifications
Thread Adapter
ceramus™ classic and EM-dispenser
45 mm
PTFE
1 Ea.
32 mm