Learn More
Fisherbrand™ PTFE Total Immersion Temperature Probe
Fisherbrand™ PTFE Total Immersion Temperature Probe are suitable for use in autoclaves and most corrosive liquids.
Brand:  Fisherbrand™ 500.050.1.2
 View more versions of this product
Product Code. 16206071
  • 70.20 € / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sensor - PRT100 Class A
  • Cable - 4 Core PFA insulated with white PFA oversheath
  • Cable fused to body
  • Supplied without plug
  • Inert and non contaminating
  • Pressure Resistant
  • Temperature Range -180°C to +280°C
Specifications
Specifications
Fisherbrand™ PTFE Total Immersion Temperature Probe
1
-180°C to +280°C
2 m (Cable)
PTFE
PRT100 Class A
50 x 5 mm
5 mm