Learn More
Thermo Scientific Chemicals PTFE Tweezers-Forceps
Totally inert and non-contaminating
Brand:  Thermo Scientific Chemicals 038261.KT
 View more versions of this product
Product Code. 15416195
  • 140.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • PTFE
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tweezers-Forceps
Square
200 mm
PTFE
7.8 in.