missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ PTFE Ultrapure Barrel Pump with Discharge Tube

The PTFE Ultrapure Barrel Pump with Discharge Tube is suitable for the transfer of ultra-pure liquids and those that attack conventional materials such as PP or PVC. The stopcock prevents vapours from escaping, and stops dripping after transfer.

795.00 € - 861.00 €

Specifications

Material PTFE
Pump Tube Length 1.2 m
Resistance Properties Chemicals
Type Barrel Pump
Applications Ultra-Pure Liquid Transfer, chemically aggressive liquids
View More Specs

Products 2
Product Code Brand Flow Rate Price Quantity & Availability  
Product Code Brand Flow Rate Price Quantity & Availability  
10042329
View Documents
Buerkle™
5606-0600
270 mL/stroke
795.00 €
Each
In Stock
to see stock.
Add to basket
 
10127921
View accessory products View Documents
Buerkle™
5606-1000
400 mL/stroke
861.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
Description

Description

  • Barrel pump with rigid discharge tube with stopcock made of PTFE
  • The barrel pump is also suitable for transferring ultra-pure liquids
  • The stopcock prevents hazardous vapours from escaping, and stops dripping after transfer
  • Immersion tube Ø 32 mm
  • All valves are made of solid PTFE/PVDF
  • Top quality - 3 years' warranty
Specifications

Specifications

PTFE
Chemicals
Ultra-Pure Liquid Transfer, chemically aggressive liquids
1.2 m
Barrel Pump
Tube diameter of 32 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.