Learn More
Duran™ PTFE Valve Pins
Brand:  Duran™ 286057404
 View more versions of this product
Product Code. 10705720
  • 22.55 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Valve Pin
PTFE
With Abdrehsicherung
Passage Size: 0 to 4mm
For Produran-valve tap