missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Vial
Thick wall construction. Fisherbrand™ PTFE Vials are ideal for storing aggressive or expensive materials.
60.45 € - 72.70 €
Specifications
Color | White |
---|---|
Material | PTFE |
For Use With (Application) | Storage of hazardous chemicals |
Graduated | No |
Temperature Range (Metric) | -200°C to +280°C |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10650283
|
Fisherbrand™
014.150 |
50 mL |
72.70 €
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
10762793
|
Fisherbrand™
014.125 |
25 mL |
60.45 €
Each |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||||
Description
- Suitable for small scale reactions at low pressure
- Tapered inner for easy removal of contents
Specifications
White | |
Storage of hazardous chemicals | |
-200°C to +280°C | |
Screw Cap | |
Certificate of Analysis (COA), Certificate of Conformance (COC) |
PTFE | |
No | |
Autoclavable | |
1 Ea. |